Assay Details
| Gene Symbol: MAP1LC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283869.1 | NP_001270798.1 | 2 - 3 | 202 | 66 | |
| XM_005592735.2 | XP_005592792.1 | 2 - 3 | 322 | 66 | ||
| GenBank mRNA | AB169714.1 | - | 2 - 3 | 207 | 66 | |
| AB170381.1 | - | 2 - 3 | 202 | 66 | ||
More Information
Set Membership: |
|

MAP1LC3B
Genomic Map