Assay Details
| Gene Symbol: EGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565754.2 | XP_005565811.1 | 2 - 3 | 323 | 71 | |
| XM_005565755.1 | XP_005565812.1 | 2 - 3 | 424 | 71 | ||
| XM_015456298.1 | XP_015311784.1 | 2 - 3 | 317 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005565755.1 transcripts.
Set Membership: |
|

EGR2
Genomic Map