Assay Details
| Gene Symbol: TXNL4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592413.2 | XP_005592470.1 | 3 - 4 | 543 | 86 | |
| XM_005592414.2 | XP_005592471.1 | 2 - 3 | 447 | 86 | ||
| XM_005592415.2 | XP_005592472.1 | 3 - 4 | 557 | 86 | ||
| XM_015442918.1 | XP_015298404.1 | 3 - 4 | 488 | 86 | ||
| XM_015442919.1 | XP_015298405.1 | 2 - 3 | 447 | 86 | ||
| XM_015442920.1 | XP_015298406.1 | 2 - 3 | 335 | 86 | ||
| XM_015442921.1 | XP_015298407.1 | 2 - 3 | 408 | 86 | ||
More Information
Set Membership: |
|

TXNL4B
Genomic Map