Assay Details
| Gene Symbol: GSTT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289959.1 | NP_001276888.1 | 113 | 64 | ||
| GenBank mRNA | KC734511.1 | - | 113 | 64 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GSTT2| Gene Symbol: GSTT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289959.1 | NP_001276888.1 | 113 | 64 | ||
| GenBank mRNA | KC734511.1 | - | 113 | 64 | ||