Assay Details
| Gene Symbol: LAMTOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541490.2 | XP_005541547.1 | 1 - 2 | 235 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LAMTOR2| Gene Symbol: LAMTOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541490.2 | XP_005541547.1 | 1 - 2 | 235 | 93 | |
Set Membership: |
|