Assay Details
| Gene Symbol: MSMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556240.2 | XP_005556297.1 | 5 - 6 | 853 | 98 | |
| XM_005556242.2 | XP_005556299.1 | 4 - 5 | 520 | 98 | ||
| XM_015451005.1 | XP_015306491.1 | 6 - 7 | 901 | 98 | ||
| XM_015451006.1 | XP_015306492.1 | 4 - 5 | 713 | 98 | ||
| GenBank mRNA | AB169840.1 | - | 5 - 6 | 755 | 98 | |
| AB170578.1 | - | 5 - 6 | 824 | 98 | ||
More Information
Set Membership: |
|

MSMO1
Genomic Map