Assay Details
| Gene Symbol: TCF21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551886.2 | XP_005551943.1 | 1 - 2 | 832 | 81 | |
| XM_005551887.2 | XP_005551944.1 | 1 - 2 | 1739 | 81 | ||
More Information
Set Membership: |
|

TCF21
Genomic Map