Assay Details
| Gene Symbol: KTI12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543376.1 | XP_005543433.1 | 1 | 1433 | 65 | |
| GenBank mRNA | AB172572.1 | - | 1 | 510 | 65 | |
| AB172573.1 | - | 1 | 510 | 65 | ||
More Information
Set Membership: |
|

KTI12
Genomic Map