Assay Details
| Gene Symbol: WDR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547064.2 | XP_005547121.1 | 5 - 6 | 3275 | 64 | |
| XM_005547065.2 | XP_005547122.1 | 5 - 6 | 3782 | 64 | ||
More Information
Set Membership: |
|

WDR6
Genomic Map