Assay Details
| Gene Symbol: RNF139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564039.2 | XP_005564096.1 | 1 - 2 | 521 | 106 | |
| GenBank mRNA | AB168971.1 | - | 1 - 2 | 531 | 106 | |
More Information
Set Membership: |
|

RNF139
Genomic Map