Assay Details
| Gene Symbol: PSMG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015449955.1 | XP_015305441.1 | 1 - 2 | 253 | 57 | |
| XM_015449956.1 | XP_015305442.1 | 1 - 2 | 253 | 57 | ||
| XM_015449957.1 | XP_015305443.1 | 1 - 2 | 253 | 57 | ||
| XM_015449958.1 | XP_015305444.1 | 1 - 2 | 280 | 57 | ||
| XM_015449959.1 | XP_015305445.1 | 1 - 2 | 280 | 57 | ||
| XM_015449960.1 | XP_015305446.1 | 1 - 2 | 280 | 57 | ||
More Information
Set Membership: |
|

PSMG4
Genomic Map