Assay Details
| Gene Symbol: MAPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586689.2 | XP_005586746.1 | 4 - 5 | 1900 | 89 | |
| XM_005586690.2 | XP_005586747.1 | 4 - 5 | 1868 | 89 | ||
| XM_005586695.2 | XP_005586752.1 | 4 - 5 | 2930 | 89 | ||
| XM_015439942.1 | XP_015295428.1 | 5 - 6 | 2161 | 89 | ||
| XM_015439943.1 | XP_015295429.1 | 4 - 5 | 2081 | 89 | ||
| XM_015439944.1 | XP_015295430.1 | 5 - 6 | 1979 | 89 | ||
More Information
Set Membership: |
|

MAPK4
Genomic Map