Assay Details
| Gene Symbol: DUSP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594908.2 | XP_005594965.1 | 2 - 3 | 637 | 65 | |
| XM_005594909.2 | XP_005594966.1 | 2 - 3 | 578 | 65 | ||
More Information
Set Membership: |
|

DUSP9
Genomic Map