Assay Details
| Gene Symbol: OCRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015444102.1 | XP_015299588.1 | 16 - 17 | 2071 | 105 | |
| GenBank mRNA | AB179082.1 | - | 8 - 9 | 926 | 105 | |
More Information
Set Membership: |
|

OCRL
Genomic Map