Assay Details
| Gene Symbol: ANAPC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585268.2 | XP_005585325.1 | 2 - 3 | 267 | 115 | |
| XM_005585269.2 | XP_005585326.1 | 2 - 3 | 292 | 115 | ||
| XM_005585270.2 | XP_005585327.1 | 2 - 3 | 236 | 115 | ||
| XM_005585271.2 | XP_005585328.1 | 2 - 3 | 420 | 115 | ||
| XM_005585272.2 | XP_005585329.1 | 3 - 4 | 359 | 115 | ||
| XM_015437953.1 | XP_015293439.1 | 3 - 4 | 331 | 115 | ||
More Information
Set Membership: |
|

ANAPC11
Genomic Map