Assay Details
| Gene Symbol: CLDN22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556372.2 | XP_005556429.1 | 1 | 620 | 73 | |
| Gene Symbol: WWC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556373.2 | XP_005556430.1 | 24 | 8135 | 73 | |
| XM_005556375.2 | XP_005556432.1 | 24 | 8219 | 73 | ||
| XM_015451055.1 | XP_015306541.1 | 25 | 8339 | 73 | ||
| XM_015451056.1 | XP_015306542.1 | 25 | 8255 | 73 | ||
| XM_015451057.1 | XP_015306543.1 | 21 | 9219 | 73 | ||
More Information
Set Membership: |
|

CLDN22
Genomic Map