Assay Details
| Gene Symbol: VAMP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575415.2 | XP_005575472.1 | 2 - 3 | 230 | 78 | |
| XM_015433473.1 | XP_015288959.1 | 2 - 3 | 1247 | 78 | ||
More Information
Set Membership: |
|

VAMP5
Genomic Map