Assay Details
| Gene Symbol: C2CD2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579894.2 | XP_005579951.1 | 7 - 8 | 1417 | 82 | |
| XM_005579895.2 | XP_005579952.1 | 8 - 9 | 859 | 82 | ||
| XM_015435821.1 | XP_015291307.1 | 7 - 8 | 1417 | 82 | ||
| XM_015435822.1 | XP_015291308.1 | 8 - 9 | 896 | 82 | ||
| GenBank mRNA | AB174341.1 | - | 7 - 8 | 1414 | 82 | |
More Information
Set Membership: |
|

C2CD2L
Genomic Map