Assay Details
| Gene Symbol: PMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285309.1 | NP_001272238.1 | 1 - 2 | 124 | 91 | |
| GenBank mRNA | AB170703.1 | - | 1 - 2 | 129 | 91 | |
| AB174003.1 | - | 1 - 2 | 124 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PMP2| Gene Symbol: PMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285309.1 | NP_001272238.1 | 1 - 2 | 124 | 91 | |
| GenBank mRNA | AB170703.1 | - | 1 - 2 | 129 | 91 | |
| AB174003.1 | - | 1 - 2 | 124 | 91 | ||