Assay Details
| Gene Symbol: EIF2S2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283476.1 | NP_001270405.1 | 8 - 9 | 945 | 89 | |
| XM_015457649.1 | XP_015313135.1 | 8 - 9 | 944 | 89 | ||
| XM_015457650.1 | XP_015313136.1 | 8 - 9 | 1069 | 89 | ||
| GenBank mRNA | AB169578.1 | - | 8 - 9 | 948 | 89 | |
| AB170907.1 | - | 8 - 9 | 945 | 89 | ||
More Information
Set Membership: |
|

EIF2S2
Genomic Map