Assay Details
| Gene Symbol: FZD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579302.2 | XP_005579359.1 | 1 - 2 | 604 | 90 | |
| XM_005579303.2 | XP_005579360.1 | 1 - 2 | 598 | 90 | ||
| XM_005579304.2 | XP_005579361.1 | 1 - 2 | 598 | 90 | ||
More Information
Set Membership: |
|

FZD4
Genomic Map