Assay Details
| Gene Symbol: ZHX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564020.2 | XP_005564077.1 | 3 - 4 | 3240 | 111 | |
| XM_005564021.2 | XP_005564078.1 | 3 - 4 | 3335 | 111 | ||
| XM_005564022.2 | XP_005564079.1 | 3 - 4 | 3280 | 111 | ||
More Information
Set Membership: |
|

ZHX1
Genomic Map