Assay Details
| Gene Symbol: ASIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583416.2 | XP_005583473.1 | 5 - 6 | 1855 | 79 | |
| XM_005583418.2 | XP_005583475.1 | 5 - 6 | 1038 | 79 | ||
| GenBank mRNA | AB220477.1 | - | 5 - 6 | 712 | 79 | |
| AB220554.1 | - | 5 - 6 | 1228 | 79 | ||
More Information
Set Membership: |
|

ASIC2
Genomic Map