Assay Details
| Gene Symbol: UBE2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574770.2 | XP_005574827.1 | 6 - 7 | 546 | 87 | |
| XM_005574771.2 | XP_005574828.1 | 6 - 7 | 546 | 87 | ||
| XM_015433034.1 | XP_015288520.1 | 6 - 7 | 660 | 87 | ||
More Information
Set Membership: |
|

UBE2F
Genomic Map