Assay Details
| Gene Symbol: ARSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283217.1 | NP_001270146.1 | 10 - 11 | 1756 | 76 | |
| XM_015443293.1 | XP_015298779.1 | 9 - 10 | 2022 | 76 | ||
| GenBank mRNA | AB125152.1 | - | 10 - 11 | 1756 | 76 | |
More Information
Set Membership: |
|

ARSL
Genomic Map