Assay Details
| Gene Symbol: SLC10A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283323.1 | NP_001270252.1 | 4 - 5 | 1077 | 72 | |
| GenBank mRNA | AK240619.1 | - | 4 - 5 | 973 | 72 | |
| AK240620.1 | - | 4 - 5 | 1078 | 72 | ||
| KP453714.1 | - | 4 - 5 | 943 | 72 | ||
More Information
Set Membership: |
|

SLC10A1
Genomic Map