Assay Details
| Gene Symbol: ANKRD46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283742.1 | NP_001270671.1 | 4 - 5 | 635 | 73 | |
| XM_005563807.2 | XP_005563864.1 | 5 - 6 | 743 | 73 | ||
| XM_005563808.2 | XP_005563865.1 | 4 - 5 | 887 | 73 | ||
| XM_005563809.2 | XP_005563866.1 | 4 - 5 | 686 | 73 | ||
| XM_005563810.2 | XP_005563867.1 | 5 - 6 | 744 | 73 | ||
| XM_015454805.1 | XP_015310291.1 | 5 - 6 | 957 | 73 | ||
| XM_015454806.1 | XP_015310292.1 | 4 - 5 | 1150 | 73 | ||
| GenBank mRNA | AB172661.1 | - | 4 - 5 | 635 | 73 | |
More Information
Set Membership: |
|

ANKRD46
Genomic Map