Assay Details
| Gene Symbol: MED21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284102.1 | NP_001271031.1 | 1 - 2 | 77 | 71 | |
| GenBank mRNA | AB169149.1 | - | 1 - 2 | 77 | 71 | |
More Information
Set Membership: |
|

MED21
Genomic Map