Assay Details
| Gene Symbol: SLCO1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284540.1 | NP_001271469.1 | 13 - 14 | 1937 | 85 | |
| XM_015430176.1 | XP_015285662.1 | 16 - 17 | 2204 | 85 | ||
| GenBank mRNA | JX866725.1 | - | 13 - 14 | 1937 | 85 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				

SLCO1B1
Genomic Map