Assay Details
| Gene Symbol: ACTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285025.1 | NP_001271954.1 | 1 - 2 | 82 | 54 | |
| GenBank mRNA | AB169683.1 | - | 1 - 2 | 202 | 54 | |
| AB170391.1 | - | 1 - 2 | 82 | 54 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001285025.1 transcripts.
Set Membership: |
|

ACTB
Genomic Map