Assay Details
| Gene Symbol: KLRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285324.1 | NP_001272253.1 | 5 - 6 | 592 | 95 | |
| XM_015430177.1 | XP_015285663.1 | 4 - 5 | 1114 | 95 | ||
| GenBank mRNA | AJ426430.1 | - | 5 - 6 | 592 | 95 | |
More Information
Set Membership: |
|

KLRF1
Genomic Map