Assay Details
| Gene Symbol: TICAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285821.1 | NP_001272750.1 | 1 | 521 | 79 | |
| XM_015451416.1 | XP_015306902.1 | 2 | 969 | 79 | ||
| GenBank mRNA | AB446496.1 | - | 1 | 521 | 79 | |
More Information
Set Membership: |
|

TICAM2
Genomic Map