Assay Details
| Gene Symbol: CELA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287702.1 | NP_001274631.1 | 2 - 3 | 138 | 70 | |
| GenBank mRNA | X91400.1 | - | 138 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CELA1| Gene Symbol: CELA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287702.1 | NP_001274631.1 | 2 - 3 | 138 | 70 | |
| GenBank mRNA | X91400.1 | - | 138 | 70 | ||
Set Membership: |
|