Assay Details
| Gene Symbol: LOC102137053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287711.1 | NP_001274640.1 | 3 - 4 | 247 | 65 | |
| XM_005567098.2 | XP_005567155.1 | 3 - 4 | 464 | 65 | ||
| GenBank mRNA | AB169480.1 | - | 3 - 4 | 247 | 65 | |
| AB179491.1 | - | 3 - 4 | 247 | 65 | ||
More Information
Set Membership: |
|

LOC102137053
Genomic Map