Assay Details
| Gene Symbol: OPN1SW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283204.1 | NP_001270133.1 | 4 - 5 | 930 | 82 | |
| GenBank mRNA | AF158977.1 | - | 4 - 5 | 930 | 82 | |
More Information
Set Membership: |
|

OPN1SW
Genomic Map