Assay Details
| Gene Symbol: CCNH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284583.1 | NP_001271512.1 | 4 - 5 | 619 | 81 | |
| XM_015451356.1 | XP_015306842.1 | 4 - 5 | 762 | 81 | ||
| XM_015451357.1 | XP_015306843.1 | 4 - 5 | 762 | 81 | ||
| XM_015451358.1 | XP_015306844.1 | 4 - 5 | 762 | 81 | ||
| XM_015451359.1 | XP_015306845.1 | 4 - 5 | 762 | 81 | ||
| XM_015451360.1 | XP_015306846.1 | 4 - 5 | 762 | 81 | ||
| GenBank mRNA | AB168718.1 | - | 4 - 5 | 619 | 81 | |
| AB170747.1 | - | 4 - 5 | 636 | 81 | ||
More Information
Set Membership: |
|

CCNH
Genomic Map