Assay Details
| Gene Symbol: MAGEB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284771.1 | NP_001271700.1 | 2 - 3 | 1196 | 132 | |
| GenBank mRNA | AB168198.1 | - | 2 - 3 | 1196 | 132 | |
More Information
Set Membership: |
|

MAGEB18
Genomic Map