Assay Details
| Gene Symbol: IL7R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284908.1 | NP_001271837.1 | 1 - 2 | 83 | 65 | |
| GenBank mRNA | DQ226543.1 | - | 1 - 2 | 83 | 65 | |
More Information
Set Membership: |
|

IL7R
Genomic Map