Assay Details
| Gene Symbol: PGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283759.1 | NP_001270688.1 | 2 - 3 | 354 | 64 | |
| XM_005585132.2 | XP_005585189.1 | 2 - 3 | 377 | 64 | ||
| XM_015437823.1 | XP_015293309.1 | 2 - 3 | 377 | 64 | ||
| GenBank mRNA | AB171690.1 | - | 2 - 3 | 356 | 64 | |
More Information
Set Membership: |
|

PGS1
Genomic Map