Assay Details
| Gene Symbol: VAPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283801.1 | NP_001270730.1 | 5 - 6 | 772 | 74 | |
| XM_005587188.2 | XP_005587245.1 | 6 - 7 | 1331 | 74 | ||
| XM_015439782.1 | XP_015295268.1 | 6 - 7 | 1204 | 74 | ||
| XM_015439783.1 | XP_015295269.1 | 5 - 6 | 1042 | 74 | ||
| GenBank mRNA | AB170744.1 | - | 5 - 6 | 772 | 74 | |
More Information
Set Membership: |
|

VAPA
Genomic Map