Assay Details
| Gene Symbol: SEPTIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283908.1 | NP_001270837.1 | 12 - 13 | 1515 | 83 | |
| XM_005583840.2 | XP_005583897.1 | 11 - 12 | 1436 | 83 | ||
| XM_005583841.2 | XP_005583898.1 | 10 - 11 | 2375 | 83 | ||
| XM_005583842.2 | XP_005583899.1 | 11 - 12 | 1534 | 83 | ||
| XM_015437824.1 | XP_015293310.1 | 13 - 14 | 3107 | 83 | ||
| GenBank mRNA | AB170533.1 | - | 12 - 13 | 1553 | 83 | |
| AB174295.1 | - | 12 - 13 | 1515 | 83 | ||
More Information
Set Membership: |
|

SEPTIN4
Genomic Map