Assay Details
| Gene Symbol: FAM207A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284169.1 | NP_001271098.1 | 3 - 4 | 315 | 79 | |
| XM_015446608.1 | XP_015302094.1 | 3 - 4 | 417 | 79 | ||
| XM_015446609.1 | XP_015302095.1 | 3 - 4 | 417 | 79 | ||
| XM_015446610.1 | XP_015302096.1 | 3 - 4 | 417 | 79 | ||
| XM_015446611.1 | XP_015302097.1 | 3 - 4 | 314 | 79 | ||
| GenBank mRNA | AB168895.1 | - | 3 - 4 | 315 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015446611.1 transcripts.
Set Membership: |
|

FAM207A
Genomic Map