Assay Details
| Gene Symbol: YAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284923.1 | NP_001271852.1 | 3 - 4 | 374 | 144 | |
| XM_005570594.2 | XP_005570651.1 | 4 - 5 | 488 | 144 | ||
| XM_005570596.2 | XP_005570653.1 | 2 - 3 | 2625 | 144 | ||
| GenBank mRNA | AB172052.1 | - | 3 - 4 | 374 | 144 | |
More Information
Set Membership: |
|

YAF2
Genomic Map