Assay Details
| Gene Symbol: CDC37L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285361.1 | NP_001272290.1 | 5 - 6 | 944 | 91 | |
| GenBank mRNA | AB170833.1 | - | 5 - 6 | 944 | 91 | |
More Information
Set Membership: |
|

CDC37L1
Genomic Map