Assay Details
| Gene Symbol: LDLRAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283171.1 | NP_001270100.1 | 3 - 4 | 279 | 91 | |
| XM_005587260.2 | XP_005587317.1 | 6 - 7 | 4253 | 91 | ||
| XM_005587261.2 | XP_005587318.1 | 6 - 7 | 1161 | 91 | ||
| XM_005587262.2 | XP_005587319.1 | 5 - 6 | 1949 | 91 | ||
| XM_005587263.2 | XP_005587320.1 | 5 - 6 | 2289 | 91 | ||
| XM_005587265.2 | XP_005587322.1 | 5 - 6 | 1821 | 91 | ||
| XM_015439784.1 | XP_015295270.1 | 6 - 7 | 2341 | 91 | ||
| XM_015439785.1 | XP_015295271.1 | 6 - 7 | 891 | 91 | ||
| XM_015439786.1 | XP_015295272.1 | 4 - 5 | 2567 | 91 | ||
| GenBank mRNA | AB171923.1 | - | 3 - 4 | 279 | 91 | |
More Information
Set Membership: |
|

LDLRAD4
Genomic Map