Assay Details
| Gene Symbol: INKA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283550.1 | NP_001270479.1 | 1 - 2 | 684 | 116 | |
| GenBank mRNA | AB170785.1 | - | 1 - 2 | 684 | 116 | |
More Information
Set Membership: |
|

INKA1
Genomic Map