Assay Details
| Gene Symbol: MYD88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283666.1 | NP_001270595.1 | 5 - 6 | 2276 | 81 | |
| GenBank mRNA | AB172771.1 | - | 5 - 6 | 2276 | 81 | |
More Information
Set Membership: |
|

MYD88
Genomic Map