Assay Details
| Gene Symbol: AGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284672.1 | NP_001271601.1 | 7 - 8 | 837 | 70 | |
| XM_005556356.2 | XP_005556413.1 | 7 - 8 | 1096 | 70 | ||
| XM_005556357.2 | XP_005556414.1 | 7 - 8 | 932 | 70 | ||
| XM_015450529.1 | XP_015306015.1 | 7 - 8 | 932 | 70 | ||
| GenBank mRNA | AB169261.1 | - | 7 - 8 | 837 | 70 | |
More Information
Set Membership: |
|

AGA
Genomic Map