Assay Details
| Gene Symbol: FBXO25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284870.1 | NP_001271799.1 | 7 - 8 | 710 | 78 | |
| XM_005562490.2 | XP_005562547.1 | 7 - 8 | 726 | 78 | ||
| XM_005562494.2 | XP_005562551.1 | 6 - 7 | 730 | 78 | ||
| XM_005562496.2 | XP_005562553.1 | 6 - 7 | 730 | 78 | ||
| XM_015454349.1 | XP_015309835.1 | 6 - 7 | 498 | 78 | ||
| XM_015454350.1 | XP_015309836.1 | 6 - 7 | 497 | 78 | ||
| XM_015454352.1 | XP_015309838.1 | 6 - 7 | 498 | 78 | ||
| XM_015454353.1 | XP_015309839.1 | 3 - 4 | 268 | 78 | ||
| GenBank mRNA | AB179395.1 | - | 7 - 8 | 710 | 78 | |
More Information
Set Membership: |
|

FBXO25
Genomic Map