Assay Details
| Gene Symbol: PPP3R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284973.1 | NP_001271902.1 | 1 | 218 | 73 | |
| GenBank mRNA | AB168512.1 | - | 1 | 218 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP3R2| Gene Symbol: PPP3R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284973.1 | NP_001271902.1 | 1 | 218 | 73 | |
| GenBank mRNA | AB168512.1 | - | 1 | 218 | 73 | |
Set Membership: |
|